Learn More
Corning™ CoolBox™ XT Systems Available on GSA/VA Contract for Federal Government customers only.
Provide sample cooling or freezing without ice, electricity, or batteries.
Manufacturer:  Corning™ 432033
 View more versions of this product
Catalog No. 07-210-033
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Accommodates variety of sample formats and temperatures
  • Modular design holds wide array of sample tubes and plates
  • Enables cooling up to 16 hours or freezing up to 8 hours with one Corning XT Cooling Core
  • Reduces risk of ice contamination
  • Available in single- or double-capacity systems
  • Internal cooling or freezing cores provide hours of 0.5°C to 4°C cooling or −20°C to 0°C sample freezing
  • Can be used with dry ice
  • Compatible with Corning CoolRack™ and Corning CoolSink™ thermo-conductive sample modules
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Benchtop cooling system
Enables cooling for 16 hours, freezing for 8 hours
CoolBox 2XT PCR system
Green
CoolBox 2XT, CoolRack XT M24, and CoolRack PCR96