Learn More
Corning™ CoolBox™ XT Systems
Provide sample cooling or freezing without ice, electricity, or batteries.
Manufacturer:  Corning™ 432032
 View more versions of this product
Catalog No. 07-210-032
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Accommodates variety of sample formats and temperatures
  • Modular design holds wide array of sample tubes and plates
  • Enables cooling up to 16 hours or freezing up to 8 hours with one Corning XT Cooling Core
  • Reduces risk of ice contamination
  • Available in single- or double-capacity systems
  • Internal cooling or freezing cores provide hours of 0.5°C to 4°C cooling or −20°C to 0°C sample freezing
  • Can be used with dry ice
  • Compatible with Corning CoolRack™ and Corning CoolSink™ thermo-conductive sample modules
Specifications
Specifications
Benchtop cooling system
Enables cooling for 16 hours, freezing for 8 hours
CoolBox XT PCR Strip system
Green
CoolBox XT and CoolRack XT M-PCR