Learn More
Corning™ CoolCell™ SV2 and SV10 Freezing Containers
Ensure standardized controlled-rate of -1°C/minute cell freezing in a -80°C freezer without alcohol or any fluids. Utilizes thermo-conductive alloy core and highly-insulative outer material to control rate of heat removal and provide reproducible cell cryopreservation.
$756.00
Specifications
Color | Purple |
---|---|
Shape | Round |
Type | Freezer Box |
Quantity | 1/Pk., 1/Cs. |
Weight (Metric) | 0.5 kg |
Catalog Number | Mfr. No. | Type | Holds | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Holds | Price | Quantity & Availability | ||||
07-210-010
|
Corning™
432010 |
Freezer Box | 12 (2 mL Serum Vials) |
Case for $756.00
|
|
||||
07-210-011
|
Corning™
432011 |
Freezer Box | 6 (10 mL Serum Vials) |
Case for $756.00
|
|
||||
Description
Corning CoolCell, in combination with a -80°C freezer, will provide alcohol-free freezing at the rate of -1°C/minute that is ideal for cryopreservation of most cells and cell lines. Using a combination of uniform-density cross-linked polyethylene foam, a solid state core, and radial vial symmetry, freezing profiles are consistent and reproducible. The foam is non-absorbent and will impose negligible change in the freezer environment; thereby protecting nearby frozen samples. The removal of vials is accomplished by tipping the device upside down so the vials slide out.
Features: Unique features of controlled-rate freezing with Corning™ CoolCell™ include:
- Ease of use
- Alcohol and fluid-free freezing
- Lower cost of use than alcohol-based devices
- High cell recovery and cell viability
- Reproducibility
- Simple, consistent way to standardize controlled-rate freezing
Specifications
Purple | |
Freezer Box | |
0.5 kg |
Round | |
1/Pk., 1/Cs. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.