Learn More
Corning™ CoolCell™ Stem Cell Cryopreservation System
Alcohol-free cell freezing system that controls the rate of freezing to -17°C/minute when placed in a -80°C freezer.
Manufacturer:  Corning™ 432012
 View more versions of this product
Catalog No. 07-210-012
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Includes XT Starter Benchtop Cooler, Corning CoolRack and CoolCell Cell freezing container for 6 injectable vials
  • Color: Purple
Specifications
Specifications
Includes XT Starter Benchtop Cooler, Corning CoolRack SV2 and CoolCell SV2 Cell Freezing Container for 12 Injectable Vials
Cryopreservation system
CoolCell SV2