Thermo Scientific™ Accessories for MaxQ™ 7000 Water Bath Shakers Available on GSA/VA Contract for Federal Government customers only.
Optimize the Accessories for Thermo Scientific™ MaxQ™ 7000 Water Bath Shakers to increase the versatility of the MaxQ 7000.
Manufacturer: Thermo Scientific™ SHK700037
 View more versions of this product
Catalog No. 11-675-464
  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
    Specifications
    Type Cooling Coil
    For Use With (Equipment) MaxQ™ 7000 Shaker
    Description Cooling coil for near-ambient temperature and below (requires a circulating chiller)
    Put Thermo Scientific™ Accessories for MaxQ™ 7000 Water Bath Shakers to use to increase the utility of MaxQ 7000 Water Bath Shakers. With exceptional innovation, reliability and accuracy, Thermo Scientific MaxQ 7000 Shakers, Platforms and accessories are the optimal choice for your busy lab.
  • Description & Specifications
Related Products
Product Content Feedback