Learn More
Corning™ CoolRack™ M-PF Conical Tube Modules
BIOPHARMA
Catalog No. 07210046
Click to view available options
Holds:
15 x 1.5 mL conical tubes
30 x 1.5 mL Conical Tubes
30 x 500μL Conical Tubes
Design:
M15-PF
M30-PF
Ensures ±0.1°C temperature uniformity across all tubes when cooling, snap freezing, heating or thawing.
Corning CoolRack thermo-conductive microfuge tube modules eliminate variability which originates from tubes placed directly into ice, dry ice, alcohol baths, water baths and other temperature sources. Place the CoolRack module directly onto a temperature source between -196°C to >100°C and it will rapidly adapt to that temperature.
  • Ensures ± 0.1°C temperature uniformity across all tubes when cooling, snap freezing, heating or thawing.
  • All CoolRack modules may be autoclaved, high heat sterilized or decontaminated with bleach, alcohol or other disinfectants or lab detergents.
  • Suggested applications include cooling reagents such as restriction enzymes, dNTPs and antibodies, alcohol-free dry ice snap freezing of tissue, virus and bacteria samples and bench top cryogenic tube sorting in liquid nitrogen.
TRUSTED_SUSTAINABILITY
Specifications
Holds 30 x 500μL Conical Tubes
Design M30-PF
Type Conical Tube Thermoconductive Module
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.