Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Coomassie Brilliant Blue R-250, Spectrum™

Click to view available options
Quantity:
25 g
Packaging:
Amber Glass Bottle
Drum
Poly Pail
6104-59-2
C45H44N3NaO7S2
825.97
HAEGGLCXLFWTBE-UHFFFAOYSA-M
sodium {4-[(4-ethoxyphenyl)amino]phenyl}bis(4-{ethyl[(3-sulfonatophenyl)methyl]amino}phenyl)methylium
[Na+].CCOC1=CC=C(NC2=CC=C(C=C2)[C+](C2=CC=C(C=C2)N(CC)CC2=CC=CC(=C2)S([O-])(=O)=O)C2=CC=C(C=C2)N(CC)CC2=CC=CC(=C2)S([O-])(=O)=O)C=C1
6104-59-2
C45H44N3NaO7S2
825.97
HAEGGLCXLFWTBE-UHFFFAOYSA-M
sodium {4-[(4-ethoxyphenyl)amino]phenyl}bis(4-{ethyl[(3-sulfonatophenyl)methyl]amino}phenyl)methylium
[Na+].CCOC1=CC=C(NC2=CC=C(C=C2)[C+](C2=CC=C(C=C2)N(CC)CC2=CC=CC(=C2)S([O-])(=O)=O)C2=CC=C(C=C2)N(CC)CC2=CC=CC(=C2)S([O-])(=O)=O)C=C1

CO131, 6104-59-2, C46H44N3O7S2Na

Spectrum™ Coomassie Brilliant Blue R-250
CAS Max % 1
Packaging Amber Glass Bottle
Quantity 25 g