Learn More
CoorsTek™ Porous Cups
SCI_ED
Supplier:  CoorsTek™ 60493/EMD
 View more versions of this product
Catalog No. S43345
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
Description
Description
These unglazed porcelain cylinders are designed for use with voltaic cells. Available in two popular sizes.
Specifications
Specifications
Voltaic Cell
For use in chemical processing, pharmaceutical, mechanical, and electronic applications
900°C to 1400°C
2.9 in.
1.49 in.
Porcelain, Ceramic
1652°F to 284°F
Porous Cup
7.5 cm
3.8 cm
Product Suggestions
Product Suggestions