Learn More
Cole-Parmer Porcelain Mortar, 50 mL, pack of 1  DFS Item
Manufacturer:  Cole-Parmer 1782000
CoorsTek 60310 Porcelain Mortar, 50 mL, pack of 1, Mortars are glazed all over except for grinding surfaces and the outside bottom. Features include lip and pour spout. Mortar and pestle are sold as separate pieces. Autoclavable.
Catalog No. NC1278119