Learn More
CoorsTek™ Aluminum Oxide Mortars and Pestles
Extra hardness and abrasion resistance compared to agate
$17.36 - $1248.00
Save up to 75%
Specifications
Material Alumina
Product Type Mortar
Finish Glazed outside
For Use With (Application) Used for elemental analysis, basic laboratory purposes and sample preparation and emulsifications
View More Specs
Products 6
Catalog Number Mfr. No. Capacity Diameter (English) Outer Height (English) Price Quantity & Availability  
Catalog Number Mfr. No. Capacity Diameter (English) Outer Height (English) Price Quantity & Availability  
12-948-5G
Accessories View Documents
CoorsTek™
60373
4.05 oz., 120mL 4.72 in. 2.24 in.
Each for $1,248.00
Add to cart
12-948A
Accessories View Documents
CoorsTek™
60335
0.05 oz., 1.5mL 1.37 in. 0.7 in.
Each for $400.00
Case of 4 EA for $1,368.00
Add to cart
12-948B
Accessories View Documents
CoorsTek™
60358
0.23 oz., 7mL 1.96 in. 1.02 in.
Each for $17.36
Save 75% Reg : $70.00
Case of 3 EA for $52.09
Save 95% Reg : $1,106.00
Add to cart
12-948C
Accessories View Documents
CoorsTek™
60361
0.5 oz., 15mL 2.55 in. 1.22 in.
Case of 2 for $904.00
Add to cart
12-948D
Accessories View Documents
CoorsTek™
60364
1.01 oz., 30mL 2.95 in. 1.45 in.
Each for $574.00
Add to cart
12-948F
Accessories View Documents
CoorsTek™
60370
2.70 oz., 80mL 3.93 in. 1.85 in.
Each for $987.00
Add to cart
Description
Description
Extra hardness and abrasion resistance compared to agate
  • Essentially inert
  • Outside glazed
Order Info
Without pestles; order separately.
Specifications
Specifications
Alumina
Mortar
Glazed outside
Used for elemental analysis, basic laboratory purposes and sample preparation and emulsifications