Learn More
CoorsTek™ Aluminum Oxide Pestles for Aluminum Oxide Mortars
Aluminum Oxide Pestles
Manufacturer:  CoorsTek™ 60374
 View more versions of this product
Catalog No. 12-948-5H
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Pestle
120mL Alumina mortar
For elemental analysis, basic laboratory purposes and sample preparation and emulsifications
4.60 in.
Alumina
Glazed completely except grinding surface
11.7cm