Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CoorsTek™ High-Form Porcelain Crucibles

Withstand temperatures up to 1150°C (2102°F), not to exceed 200°C/hour

Manufacturer:  CoorsTek™ 60108

 View more versions of this product

Catalog No. 07-965F



Add to cart

Description

Description

  • Glazed inside and out except for outside bottom surface and rim
  • Covers sold separately

Specifications

Specifications

50mm
Porcelain
40mL
40mm
1150°C
Glazed
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit