Learn More
CoorsTek™ Porcelain Combustion Boats
AD-998 High-Alumina labware
Manufacturer:  CoorsTek™ 65563
 View more versions of this product
Catalog No. 08-226-13C
Add to cart
Specifications
Specifications
0.16 oz.
0.16 oz., 5mL
For refractory metals such as molybdenum, platinum, rhodium, tungsten, tantalum and iridium
10mm
70mm
0.55 in.
Combustion Boat
5mL
2.75 x 0.55 x 0.39 in. (70 x 14 x 10mm)
0.39 in.
2.75 in.
Alumina, Porcelain
14mm