Learn More
CoorsTek™ Porcelain Crucibles with Filtering Discs
Disc permanently fused into crucible walls
Products 1
Catalog Number Mfr. No. Porosity Price Quantity & Availability  
Catalog Number Mfr. No. Porosity Price Quantity & Availability  
08-226-10A
View Documents
CoorsTek™
60525
Very Fine This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
Disc permanently fused into crucible walls
  • Will not disintegrate when in contact with moderately alkaline solutions or acids (except HF)
  • Stable porosity and flowrate
Order Info
Height: 1.687 in. (44mm)