Learn More
CoorsTek™ Porcelain Desiccator Plates
One large center hole
Manufacturer:  CoorsTek™ 60455
 View more versions of this product
Catalog No. 08-641B
Add to cart
Description
Description
  • Without feet
  • Glazed on one side
Specifications
Specifications
19cm
Porcelain
Desiccator
7.48 in.
Desiccator plate with 5mm perforations