Learn More
CoorsTek™ Porcelain Evaporating Dishes
Glazed except on rim for easy cleaning
$18.06 - $833.50
Save up to 75%
Specifications
Type Evaporating Dish
Interior Finish Glazed
Material Porcelain
View More Specs
Products 5
Catalog Number Mfr. No. Capacity (Metric) Diameter (Metric) Height (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Diameter (Metric) Height (Metric) Price Quantity & Availability  
08-690K
View Documents
CoorsTek™
60205
525 mL 162 mm 51 mm
Each for $18.06
Save 75% Reg : $72.23
Case of 12 EA for $216.67
Save 96% Reg : $4,910.00
Add to cart
08-690Q
View Documents
CoorsTek™
60210
3250 mL 305 mm 95 mm
Each for $833.50
Case of 3 EA for $2,500.00
Add to cart
08-690E
View Documents
CoorsTek™
60200
120 mL 94 mm 42 mm
Each for $72.80
Case of 24 EA for $1,183.00
Add to cart
08-690J
View Documents
CoorsTek™
60204
385 mL 145 mm 48 mm
Each for $296.00
Case of 12 EA for $3,550.00
Add to cart
08-690P
View Documents
CoorsTek™
60209
2100 mL 265 mm 80 mm
Each for $792.00
Case of 4 EA for $3,180.00
Add to cart
Description
Description
  • Round bottoms
  • With pour-out spouts
Specifications
Specifications
Evaporating Dish
Glazed
Porcelain