Learn More
CoorsTek™ Porcelain Gooch Filtering Crucibles
Glazed inside and out except for outside bottom surface
Manufacturer:  CoorsTek™ 60153
 View more versions of this product
Catalog No. 08-215-1
Add to cart
Description
Description
  • Perforated bottoms
  • Perforations, 0.013 in. dia. (0.7mm) in 13 to 40mL size; 0.046 in. dia. (1mm) in 130mL size
  • Covers sold separately
Specifications
Specifications
130mL
Glazed
Gooch Filtering Crucible
55mm
For total suspended solids determination
Porcelain