Learn More
CoorsTek™ Porcelain Spoons and Spatulas
Combination spatula and spoon
Manufacturer:  CoorsTek™ 60480
 View more versions of this product
Catalog No. 14-430B
Add to cart
Description
Description
Porcelain avoids scratching delicate surfaces
Specifications
Specifications
0.016 oz.
Spatula
Prevents scratching of delicate surfaces
0.016 oz., 0.5mL
1150°C
16.5cm
0.5mL
Glazed
6.5 in.
Spoon on one end and a spatula on the other
Porcelain