Learn More
CoorsTek™ Porcelain Spoons and Spatulas
Combination spatula and spoon
Manufacturer:  CoorsTek™ 60481
 View more versions of this product
Catalog No. 14-345C
Add to cart
Description
Description
Porcelain avoids scratching delicate surfaces
Specifications
Specifications
0.033 oz.
Spatula
Prevents scratching of delicate surfaces
0.033 oz., 1mL
1150°C
19.5cm
1mL
Glazed
7.62 in.
Spoon on one end and a spatula on the other
Porcelain