Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
COP1 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548171
Description
Caspase inhibitor. Acts as a regulator of procaspase- 1/CASP1 activation implicated in the regulation of the proteolytic maturation of pro-interleukin-1 beta (IL1B) and its release during inflammation. Inhibits the release of IL1B in response to LPS in monocytes. Also induces NF-kappa-B activation during the pro- inflammatory cytokine response. Also able to inhibit CASP1- mediated neuronal cell death, TNF-alpha, hypoxia-, UV-, and staurosporine-mediated cell death but not ER stress-mediated cell death. Acts by preventing activation of caspases CASP1 and CASP4, possibly by preventing the interaction between CASP1 and RIPK2.
Specifications
COP1 | |
Polyclonal | |
Unconjugated | |
CARD16 | |
Caspase recruitment domain-containing protein 16, Caspase recruitment domain-only protein 1, CARD-only protein 1, Caspase-1 inhibitor COP, Pseudo interleukin-1 beta converting enzyme, Pseudo-ICE, Pseudo-IL1B-converting enzyme, CARD16, COP, COP1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
114769 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Western Blot | |
0.35 mg/mL | |
PBS with 0.09% sodium azide | |
Q5EG05 | |
CARD16 | |
KLH conjugated synthetic peptide between 37-65 amino acids from the N-terminal region of human COP1. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction