Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COP9 Mouse anti-Human, Alexa Fluor™ 647, Clone: OTI3E1, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271382AF647

Catalog No. N271382A647


Add to cart

Description

Description

COP9 Monoclonal antibody specifically detects COP9 in Human samples. It is validated for Western Blot.
Specifications

Specifications

COP9
OTI3E1
Western Blot
Affinity Purified, Conjugated
COPS3
Recombinant protein fragment corresponding to amino acids 224-423 of human COP9 (NP_003644) produced in E.coli.
Immunogen affinity purified
Cell Cycle and Replication, Neuronal Cell Markers, Neurotransmission, Signal Transduction
Primary
8533.0
Western Blot
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
COP9 constitutive photomorphogenic homolog subunit 3 (Arabidopsis), COP9 signalosome complex subunit 3, CSN3COP9 complex subunit 3, JAB1-containing signalosome subunit 3, SGN3COP9 (constitutive photomorphogenic, Arabidopsis, homolog) subunit 3, Signalosome subunit 3
Mouse
IgG2b
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit