Copan Diagnostics CRYOBANK™Bead System Available on GSA/VA Contract for Federal Government customers only.

Easy-to-use system for freezing bacterial cultures for later retrieval

Manufacturer: Copan Diagnostics CRYO/R

 View more versions of this product

Catalog No. 14-906-52

  Check Availability
Add to Cart

    Specifications

    Product Type Culture Tube
    Color Red
    For Use With (Application) For freezing bacterial cultures for later retrieval
    Quantity 64/Pk.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit