Copan Diagnostics SnotBuster™ Sputum Liquefying Solution
Convenient, ready-to-use system for liquefying sputum specimens
Manufacturer: Copan Diagnostics 098CE.A
 View more versions of this product
Catalog No. 23-600-521
  Check Availability
Add to Cart
    Specifications
    Includes 12 x 80mm Polypropylene screw cap tubes with internal conical shape filled with 0.5mL of SL solution and sterile Sputum dippers for Sputum sample transfer
    Closure Type Screw cap
    Closure Material Polypropylene
    Product Type Sputum Liquefying Solution
  • Description & Specifications
Related Products
Product Content Feedback