Copan Diagnostics SnotBuster™ Sputum Liquefying Solution
Convenient, ready-to-use system for liquefying sputum specimens
Manufacturer: Copan Diagnostics 095CE.A
 View more versions of this product
Catalog No. 23-600-522
  Check Availability
Add to Cart
    Specifications
    Includes Twenty 12 x 80mm Polypropylene screw cap tubes with internal conical shape filled with 1mL of SL solution and 20 sterile transfer pipets for sputum sample transfer
    Closure Type Screw cap
    Closure Material Polypropylene
    Product Type Sputum Liquefying Solution and Pipet Kit
  • Description & Specifications
Related Products
Product Content Feedback