Learn More
Copan Diagnostics SnotBuster™ Sputum Liquefying Solution
Convenient, ready-to-use system for liquefying sputum specimens
Manufacturer:  Copan Diagnostics 097CE.ABX
 View more versions of this product
Catalog No. 23-600-520
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Screw cap
Polypropylene
Sputum Liquefying Solution
Thirty 12 x 80mm Polypropylene screw cap tubes with internal conical shape filled with 1mL of SL solution