Learn More
Santa Cruz Biotechnology copGFP Control Lentiviral Particles, 200 uL
Supplier:  Santa Cruz Biotechnology SC108084
Encompass
copGFP Control Lentiviral Particles are provided as transduction-ready viral particles. Lentiviral Particles contain a copGFP coding construct for copGFP expression in mammalian cells. After transduction, cells stably expressing copGFP may be isolated via puromycin selection.
Catalog No. NC9806135
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.