Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Copine I Polyclonal Antibody, Active Motif™

Rabbit Polyclonal Antibody
Supplier: Active Motif 39101

Description
Calcium-dependent membrane-binding proteins may regulate molecular events at the interface of the cell membrane and cytoplasm. This gene encodes a calcium-dependent protein that also contains two N-terminal type II C2 domains and an integrin A domain-like sequence in the C-terminus. However, the encoded protein does not contain a predicted signal sequence or transmembrane domains. This protein has a broad tissue distribution and it may function in membrane trafficking. This gene and the gene for RNA binding motif protein 12 overlap at map location 20q11.21. Alternate splicing results in multiple transcript variants encoding different proteins.Specifications
Copine I | |
Polyclonal | |
Unconjugated | |
CPNE1 | |
1810028N16Rik; BOS_13472; Chromobindin 17; copine 1; Copine I; copine I, isoform 8; copine III; copine III, like; Copine-1; COPN1; CPN1; Cpne1; cpne3; cpne3l; MGC1142; mKIAA4108; OTTHUMP00000030775; OTTHUMP00000030776; Unknown (protein for IMAGE:8126132); wu:fb69d07; wu:fc45a04; zgc:55873 | |
Rabbit | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Conc. Not Determined | |
whole serum with 30% glycerol and 0.035% sodium azide; pH | |
Q99829 | |
CPNE1 | |
This Copine I antibody was raised against a peptide corresponding to amino acids 519-532 of human Copine I. | |
RUO | |
8904 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title