Learn More
Copine I Rabbit anti-Human, Unconjugated, Polyclonal, Active Motif™
Rabbit Polyclonal Antibody
Manufacturer:  Active MotifSupplier Diversity Partner 39102
Encompass
 View more versions of this product
Catalog No. 50-199-3876
Add to cart
Description
Description
Calcium-dependent membrane-binding proteins may regulate molecular events at the interface of the cell membrane and cytoplasm. This gene encodes a calcium-dependent protein that also contains two N-terminal type II C2 domains and an integrin A domain-like sequence in the C-terminus. However, the encoded protein does not contain a predicted signal sequence or transmembrane domains. This protein has a broad tissue distribution and it may function in membrane trafficking. This gene and the gene for RNA binding motif protein 12 overlap at map location 20q11.21. Alternate splicing results in multiple transcript variants encoding different proteins.
Specifications
Specifications
Copine I
Western Blot
Unconjugated
CPNE1
Liquid
Rabbit
IgG
RUO
Antibody
Polyclonal
Human
CPNE1
Conc. Not Determined
whole serum with 30% glycerol and 0.035% sodium azide; pH
Q99829
1810028N16Rik; BOS_13472; Chromobindin 17; copine 1; Copine I; copine I, isoform 8; copine III; copine III, like; Copine-1; COPN1; CPN1; Cpne1; cpne3; cpne3l; MGC1142; mKIAA4108; OTTHUMP00000030775; OTTHUMP00000030776; Unknown (protein for IMAGE:8126132); wu:fb69d07; wu:fc45a04; zgc:55873
This Copine I antibody was raised against a peptide corresponding to amino acids 519-532 of human Copine I.
10 μL
-20°C, Avoid Freeze/Thaw Cycles
Primary
8904