Copper, 1000µg/mL, AAS standard solution, Alfa Aesar™ Specpure™

Manufacturer: Alfa Aesar 88061AP

Quantity 500mL

 View more versions of this product

Catalog No. AA88061AP

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7697-37-2,7440-50-8,7732-18-5
    Physical Form Liquid
    Molecular Formula Cu in 5% HNO3
    MDL Number MFCD00010965
    Chemical Name or Material Copper
    Grade AAS Standard

    Specifications

    Color Clear colorless
    Physical Form Liquid
    Quantity 500mL
    DOT Information DOT Class: 8, Packaging Group: III
    EINECS Number 231-714-2
    Molecular Formula Cu in 5% HNO3
    Concentration or Composition (by Analyte or Components) Nitric acid: 5%; Copper: 0.1%; Water: 94.9%
    CAS 7732-18-5,7697-37-2,7440-50-8
    MDL Number MFCD00010965
    Health Hazard 1 H314: Causes severe skin burns and eye damage.
    Vapor Pressure 17mm Hg at 20°C
    Chemical Name or Material Copper
    Grade AAS Standard

    H314: Causes severe skin burns and eye damage.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit