Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper, 99.9998%, (trace metal basis), Thermo Scientific™

Catalog Number AC437050250
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
7440-50-8
Cu
63.55
MFCD00010965
RYGMFSIKBFXOCR-UHFFFAOYSA-N
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
23978
CHEBI:30052
copper
[Cu]
7440-50-8
Cu
63.55
MFCD00010965
RYGMFSIKBFXOCR-UHFFFAOYSA-N
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
23978
CHEBI:30052
copper
[Cu]

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

For ultra trace analysis
TRUSTED_SUSTAINABILITY
Color Brown-Rust
Melting Point 1083.0°C
Boiling Point 2595.0°C
Assay Percent Range 99.9998%
Linear Formula Cu
Quantity 25 g
Solubility Information Solubility in water: insoluble.
Formula Weight 63.54
Percent Purity 99.9998%
Grade Trace Metal Basis
Packaging Glass bottle
Form Shot
Chemical Name or Material Copper
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit