Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper, 99.9998%, (trace metal basis), Acros Organics™

Click to view available options
Quantity:
25g
125g
Packaging:
Glass bottle
7440-50-8
Cu
63.546
RYGMFSIKBFXOCR-UHFFFAOYSA-N
cuprum,cobre,cuivre,blister,cathode,bronze,powder,anode,precipitates,kupfer
23978
CHEBI:30052
copper
[Cu]
7440-50-8
Cu
63.546
RYGMFSIKBFXOCR-UHFFFAOYSA-N
cuprum,cobre,cuivre,blister,cathode,bronze,powder,anode,precipitates,kupfer
23978
CHEBI:30052
copper
[Cu]
For ultra trace analysis
Quantity 125g
Assay Percent Range 99.9998%
Linear Formula Cu
Solubility Information Solubility in water: insoluble.
Packaging Glass bottle
Formula Weight 63.54
Physical Form Shot
Percent Purity 99.9998%
Grade Trace Metal Basis
Total Trace Metal Impurities 2ppm max.
Color Brown-Rust
Melting Point 1083.0°C
Boiling Point 2595.0°C
Chemical Name or Material Copper
Show More Show Less