Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper, 99%, powder, max. 106μm (-140 mesh), ACROS Organics™

Click to view available options
Quantity:
25g
500g
2.5kg
Packaging:
Glass bottle
7440-50-8
Cu
63.546
MFCD00010965
RYGMFSIKBFXOCR-UHFFFAOYSA-N
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
23978
CHEBI:30052
copper
[Cu]
7440-50-8
Cu
63.546
MFCD00010965
RYGMFSIKBFXOCR-UHFFFAOYSA-N
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
23978
CHEBI:30052
copper
[Cu]
Quantity 2.5kg
Additional Information apparent density: 1.35g/mL
Density 8.92
Assay Percent Range 99%
Linear Formula Cu
Fieser 01,157; 02,82; 03,63; 04,102; 05,146; 07,73; 08,113; 09,122; 15,99; 16,95
Merck Index 15, 2504
Packaging Glass bottle
Formula Weight 63.54
Physical Form Powder
Percent Purity 99%
Particle Size 25 micron (average), 106 micron (maximum)
Color Undesignated
Melting Point 1083.0°C
Boiling Point 2580.0°C
Chemical Name or Material Copper, 99%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable solid.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep container tightly closed.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 231-159-6

RTECSNumber : GL5325000

TSCA : TSCA