Copper based low temperature water gas shift catalyst, HiFUEL™ W220, Alfa Aesar™

Manufacturer: Alfa Aesar 45466A1

Quantity 1kg

 View more versions of this product

Catalog No. AA45466A1

  Check Availability
Add to Cart

    Chemical Identifiers

    Physical Form Pellets, 3.1mm X 3.1mm
    Chemical Name or Material Copper based low temperature water gas shift catalyst, HiFUEL W220

    Specifications

    Physical Form Pellets, 3.1mm X 3.1mm
    Quantity 1kg
    Color Black
    DOT Information DOT Class: 9, Packaging Group: III
    Health Hazard 1 H400-H410; Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.
    Appearance Black pellets
    Chemical Name or Material Copper based low temperature water gas shift catalyst, HiFUEL W220

    H400-H410; Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit