Alfa Aesar™ Copper based medium temperature water gas shift catalyst, HiFUEL™ W230

Manufacturer: Alfa Aesar™ 45470A1

Quantity 1kg

 View more versions of this product

Catalog No. AA45470A1

  Check Availability
Add to Cart

    Chemical Identifiers

    UN Number UN3077
    Chemical Name or Material Copper based medium temperature water gas shift catalyst HiFUEL™ W230

    Specifications

    Physical Form Pellet
    Health Hazard 3 GHS P Statement
    P280h-P273-P305+P351+P338-P501a
    Wear protective gloves/clothing.
    Avoid release to the environment.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 1 Warning
    TSCA TSCA
    UN Number UN3077
    Health Hazard 2 GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Color Black
    Quantity 1kg
    Particle Size 5.2mm x 3.0mm
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
    Chemical Name or Material Copper based medium temperature water gas shift catalyst HiFUEL™ W230

    GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P280h-P273-P305+P351+P338-P501a
    Wear protective gloves/clothing.
    Avoid release to the environment.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit