Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Copper based medium temperature water gas shift catalyst, HiFUEL™ W230

Manufacturer:  Alfa Aesar™ 45470A4

Quantity 2.5kg

 View more versions of this product

Catalog No. AA45470A4


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

UN3077
Copper based medium temperature water gas shift catalyst HiFUEL™ W230

Specifications

Pellet
Black
5.2mm x 3.0mm
UN3077
2.5kg
Copper based medium temperature water gas shift catalyst HiFUEL™ W230

Safety and Handling

SDS & Safety Documents

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280h-P273-P305+P351+P338-P501a
Wear protective gloves/clothing.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit