Alfa Aesar™ Copper based methanol reforming catalyst, HiFUEL™ R120

Manufacturer: Alfa Aesar™ 45468A1

Quantity 1kg

 View more versions of this product

Catalog No. AA45468A1

  Check Availability
Add to Cart

    Chemical Identifiers

    UN Number UN3077
    Chemical Name or Material Copper based methanol reforming catalyst HiFUEL™ R120

    Specifications

    Physical Form Pellet
    Health Hazard 3 GHS P Statement
    P264-P270-P301+P312-P330-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Rinse mouth.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
    UN Number UN3077
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H302
    Harmful if swallowed.
    Color Black
    Odor Odorless
    Particle Size 5.2mm x 3.0mm
    TSCA TSCA
    Quantity 1kg
    Chemical Name or Material Copper based methanol reforming catalyst HiFUEL™ R120

    GHS H Statement
    H302
    Harmful if swallowed.

    GHS P Statement
    P264-P270-P301+P312-P330-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Rinse mouth.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit