Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Copper Beryllium foil, alloy C17200, 0.4mm (0.016 in.) thick

Manufacturer:  Alfa Aesar™ 41914GU

Quantity 50x200mm

 View more versions of this product

Catalog No. AA41914GU


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

MFCD01320550
Copper Beryllium foil, Alloy C17200, 0.4mm (0.016 in.) thick

Specifications

Cu:Be; 98.1:1.9 wt%
50x200mm
MFCD01320550
Copper Beryllium foil, Alloy C17200, 0.4mm (0.016 in.) thick
≈57.4g/75x225mm
Foil
Odorless

Safety and Handling

SDS & Safety Documents

GHS H Statement
H350-H373-H332-H317
May cause cancer.
May cause damage to organs through prolonged or repeated exposure.
Harmful if inhaled.
May cause an allergic skin reaction.

GHS P Statement
P260-P261-P280-P304+P340-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit