Alfa Aesar™ Copper Beryllium rod, alloy C17200, 9.525mm (0.375 in.) dia.

Manufacturer: Alfa Aesar™ 42483BU

Quantity 25cm

 View more versions of this product

Catalog No. AA42483BU

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    MDL Number MFCD01320550
    Chemical Name or Material Copper Beryllium rod, Alloy C17200, 9.525mm (0.375 in.) dia.

    Specifications

    Health Hazard 3 GHS P Statement
    P260-P261-P280-P304+P340-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Rod
    Odor Odorless
    Health Hazard 1 Danger
    Weight ≈149.2g/25cm
    TSCA TSCA
    MDL Number MFCD01320550
    Health Hazard 2 GHS H Statement
    H350-H373-H332-H317
    May cause cancer.
    May cause damage to organs through prolonged or repeated exposure.
    Harmful if inhaled.
    May cause an allergic skin reaction.
    Avg. Mol. Wt. or Mol. Wt. Range Cu:Be; 98.1:1.9 wt%
    Quantity 25cm
    Chemical Name or Material Copper Beryllium rod, Alloy C17200, 9.525mm (0.375 in.) dia.

    GHS H Statement
    H350-H373-H332-H317
    May cause cancer.
    May cause damage to organs through prolonged or repeated exposure.
    Harmful if inhaled.
    May cause an allergic skin reaction.

    GHS P Statement
    P260-P261-P280-P304+P340-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit