Alfa Aesar™ Copper bis(trifluoromethylsulfonyl)imide

Manufacturer: Alfa Aesar™ 4716206

Quantity 5g

 View more versions of this product

Catalog No. AA4716206

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 291300-50-0
    Linear Formula [(CF3SO2)2N]Cu
    Formula Weight 343.68
    MDL Number MFCD23380170
    UN Number UN3261
    Synonym Copper bis(trifluoromethanesulfonyl)imide; Copper bis(trifluoromethanesulfonimide)
    Chemical Name or Material Copper bis(trifluoromethylsulfonyl)imide

    Specifications

    Formula Weight 343.68
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    MDL Number MFCD23380170
    UN Number UN3261
    Color Blue
    Sensitivity Hygroscopic
    DOT Information Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
    Physical Form Solid
    Linear Formula [(CF3SO2)2N]Cu
    CAS 291300-50-0
    Quantity 5g
    Synonym Copper bis(trifluoromethanesulfonyl)imide; Copper bis(trifluoromethanesulfonimide)
    Health Hazard 1 Danger
    Chemical Name or Material Copper bis(trifluoromethylsulfonyl)imide

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit