Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper flake, -325 mesh, 99% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 4507336

Quantity 500g

 View more versions of this product

Catalog No. AA4507336

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

7440-50-8
99% (Metals basis)
63.55
Copper
Flakes
Cu
MFCD00010965

Specifications

500g
Copper colored flakes
99% (Metals basis)
63.55
Odorless
Copper colored
7440-50-8
Flakes
Cu
MFCD00010965
0 hPa
Copper

Safety and Handling

SDS & Safety Documents

H228-H319-H335-H411-H401: Flammable solid. Causes serious eye irritation. May cause respiratory irritation. Toxic to aquatic life with long lasting effects. Toxic to aquatic life.

DOTInformation : DOT Class: 4.1, Packaging Group: III

EINECSNumber : 231-159-6

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit