Copper flake, -325 mesh, 99% (metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar 45073A1

Quantity 1kg

 View more versions of this product

Catalog No. AA45073A1

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-50-8
    Physical Form Flakes
    Assay Percent Range 99% (Metals basis)
    Molecular Formula Cu
    Formula Weight 63.55
    MDL Number MFCD00010965
    Chemical Name or Material Copper

    Specifications

    Formula Weight 63.55
    Quantity 1kg
    Color Copper colored
    DOT Information DOT Class: 4.1, Packaging Group: III
    Density 8.94g/cm3
    EINECS Number 231-159-6
    Physical Form Flakes
    Assay Percent Range 99% (Metals basis)
    Molecular Formula Cu
    MDL Number MFCD00010965
    CAS 7440-50-8
    Odor Odorless
    Health Hazard 1 H228-H319-H335-H411-H401: Flammable solid. Causes serious eye irritation. May cause respiratory irritation. Toxic to aquatic life with long lasting effects. Toxic to aquatic life.
    Appearance Copper colored flakes
    Vapor Pressure 0 hPa
    Chemical Name or Material Copper

    H228-H319-H335-H411-H401: Flammable solid. Causes serious eye irritation. May cause respiratory irritation. Toxic to aquatic life with long lasting effects. Toxic to aquatic life.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit