Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Copper foil, 0.025mm (0.001 in.) thick, annealed, uncoated, 99.8% (metals basis)

For Graphene growth

Manufacturer:  Alfa Aesar™ 46365RG

Quantity 30x150cm

 View more versions of this product

Catalog No. AA46365RG


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

7440-50-8
MFCD00010965
Copper foil, 0.025mm (0.001 in.) thick, annealed, uncoated

Specifications

≈20.2g/30x30cm
7440-50-8
MFCD00010965
Copper
99.8%, (metals basis)
30x150cm
Foil
Odorless
Copper foil, 0.025mm (0.001 in.) thick, annealed, uncoated

Safety and Handling

SDS & Safety Documents

RTECSNumber : GL5325000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit