Alfa Aesar™ Copper foil, 0.025mm (0.001 in.) thick, annealed, uncoated, 99.8% (metals basis)

For Graphene growth

Manufacturer: Alfa Aesar™ 46365RF

Quantity 30x30cm

 View more versions of this product

Catalog No. AA46365RF

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-50-8
    MDL Number MFCD00010965
    Chemical Name or Material Copper foil, 0.025mm (0.001 in.) thick, annealed, uncoated

    Specifications

    MDL Number MFCD00010965
    CAS 7440-50-8
    Odor Odorless
    RTECS Number GL5325000
    Weight ≈20.2g/30x30cm
    Quantity 30x30cm
    TSCA TSCA
    Physical Form Foil
    Color Copper
    Chemical Name or Material Copper foil, 0.025mm (0.001 in.) thick, annealed, uncoated
    Assay 99.8%, (metals basis)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit