Alfa Aesar™ Copper foil, 0.1mm thick, 99.99% (metals basis), Alfa Aesar™, Puratronic™

Manufacturer: Alfa Aesar™ 42973GH

Quantity 100 x 100mm

 View more versions of this product

Catalog No. AA42973GH

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-50-8
    MDL Number MFCD00010965
    Chemical Name or Material Copper

    Specifications

    Quantity 100 x 100mm
    Density 8.94g/cm3
    EINECS Number 231-159-6
    Physical Form Foil, 0.1mm (0.004 in.) thick
    Assay Percent Range 99.9999% (Metals basis)
    MDL Number MFCD00010965
    CAS 7440-50-8
    Odor Odorless
    Appearance Copper colored foil
    Vapor Pressure 0 hPa
    Chemical Name or Material Copper

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit