Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper foil, 99.99% (metals basis), Alfa Aesar™

Click to view available options
Quantity:
50 x 50mm
50 x 250mm
50 x 1000mm
7440-50-8
Cu
63.546
MFCD00010965
RYGMFSIKBFXOCR-UHFFFAOYSA-N
cuprum,cobre,cuivre,blister,cathode,bronze,powder,anode,precipitates,kupfer
23978
CHEBI:30052
copper
[Cu]
7440-50-8
Cu
63.546
MFCD00010965
RYGMFSIKBFXOCR-UHFFFAOYSA-N
cuprum,cobre,cuivre,blister,cathode,bronze,powder,anode,precipitates,kupfer
23978
CHEBI:30052
copper
[Cu]

Form: approx. 2.2g/50x50mm

Quantity 50 x 250mm
Appearance Copper colored foil
Density 8.94g/cm3
Assay Percent Range 99.9999% (Metals basis)
Odor Odorless
Physical Form Foil, 1.0mm (0.04 in.) thick
Vapor Pressure 0 hPa
Color Undesignated
Chemical Name or Material Copper

EINECSNumber : 231-159-6

TSCA : Yes

Recommended Storage : Ambient temperatures