Copper foil, Annealed, 99.8% (metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar 13382RF

Quantity 30 x 30cm

 View more versions of this product

Catalog No. AA13382RF

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-50-8
    Physical Form ∽20.2g/30 X 30cm
    Assay Percent Range 99.8% (metals basis)
    MDL Number MFCD00010965
    Chemical Name or Material Copper foil (Annealed)

    Specifications

    Physical Form ∽20.2g/30 X 30cm
    Quantity 30 x 30cm
    EINECS Number 231-159-6
    Assay Percent Range 99.8% (metals basis)
    MDL Number MFCD00010965
    CAS 7440-50-8
    Chemical Name or Material Copper foil (Annealed)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit