Copper foil, 0.127mm (0.005 in.) thick, annealed, 99.9% (metals basis)

Manufacturer: Alfa Aesar 13380CV

Quantity 20 x 100cm

 View more versions of this product

Catalog No. AA13380CV

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-50-8
    MDL Number MFCD00010965
    Chemical Name or Material Copper foil (Annealed), 0.127mm (0.005 in.) thick, Annealed, Temper: soft

    Specifications

    Quantity 20 x 100cm
    Physical Form ∽45.5g/20X100cm, Temper: Soft
    TSCA TSCA
    CAS 7440-50-8
    Assay Percent Range 99.9% (metals basis)
    MDL Number MFCD00010965
    Chemical Name or Material Copper foil (Annealed), 0.127mm (0.005 in.) thick, Annealed, Temper: soft
    Assay 99.9%, (metals basis)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit