Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper Gluconate, FCC, 98-102%, Spectrum™

Click to view available options
Quantity:
125 g
500 g
2.5 kg
25 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
527-09-3
C12H22CuO14
453.84
OCUCCJIRFHNWBP-IYEMJOQQSA-L
copper(2+) bis((2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate)
[Cu++].OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O.OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O
527-09-3
C12H22CuO14
453.84
OCUCCJIRFHNWBP-IYEMJOQQSA-L
copper(2+) bis((2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate)
[Cu++].OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O.OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O

C1329, 527-09-3, C12H22CuO14

Spectrum™ Copper Gluconate, FCC is used as a nutrient and dietary supplement. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 100%
Percent Purity 98-102%
Lead (Pb) 5 mg/kg
Packaging Poly Bottle
Quantity 500 g