Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper Gluconate, USP, 98-102%, Spectrum™

Click to view available options
Quantity:
125 g
500 g
2.5 kg
25 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
527-09-3
C12H22CuO14
453.84
OCUCCJIRFHNWBP-IYEMJOQQSA-L
copper(2+) bis((2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate)
[Cu++].OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O.OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O
527-09-3
C12H22CuO14
453.84
OCUCCJIRFHNWBP-IYEMJOQQSA-L
copper(2+) bis((2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate)
[Cu++].OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O.OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O

C1334, 527-09-3, C12H22CuO14

Spectrum™ Copper Gluconate, USP is used as a nutrient and dietary supplement. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 100%
Arsenic (As) 3 ppm
Chloride 0.07%
Percent Purity 98-102%
Lead (Pb) 25 μg/g
Packaging Poly Pail
Quantity 2.5 kg