Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper, Granular, 20 to 30 Mesh, BAKER ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
2.5kg
500g
Packaging:
Poly Bottle

Chemical Identifiers

7440-50-8
Cu
63.546
RYGMFSIKBFXOCR-UHFFFAOYSA-N
23978
CHEBI:30052

High quality chemicals for laboratory and specialized industrial use

Specific Gravity 1L = 8.94kg
Quantity 500g
CAS 7440-50-8
Molecular Formula Cu
Mesh Size 20 to 30 Mesh
InChI Key RYGMFSIKBFXOCR-UHFFFAOYSA-N
SMILES [Cu]
Packaging Poly Bottle
IUPAC Name copper
Molecular Weight (g/mol) 63.546
PubChem CID 23978
ChEBI CHEBI:30052
Formula Weight 63.55
Physical Form Granular
Chemical Name or Material Copper
Grade BAKER ANALYZED™ Reagent
Show More Show Less